3,4-Diaminoanisole

Supplier: TCI America

Synonyms: 4-Methoxy-o-phenylenediamine

M2882-1G M2882-5G
TCM2882-1GEA 77.25 USD
TCM2882-1G TCM2882-5G
3,4-Diaminoanisole
3,4-Diaminoanisole
Formula: C₇H₁₀N₂O
MDL Number: MFCD00047837
CAS Number: 102-51-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR