2-Methoxy-5-sulfamoylbenzoic acid

Supplier: TCI America

M2771-25G M2771-5G
TCM2771-25GEA 159.65 USD
TCM2771-25G TCM2771-5G
2-Methoxy-5-sulfamoylbenzoic acid
2-Methoxy-5-sulphamoylbenzoic acid
Formula: C₈H₉NO₅S
MDL Number: MFCD00129997
CAS Number: 22117-85-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR