Methanesulfonylacetic acid

Supplier: TCI America

Synonyms: 2-(Methylsulphonyl)acetic acid

M2770-1G M2770-5G
TCM2770-1GEA 43.26 USD
TCM2770-1G TCM2770-5G
Methanesulfonylacetic acid
Methanesulfonylacetic acid
Formula: C₃H₆O₄S
MW: 138.14 g/mol
MDL Number: MFCD00051843
CAS Number: 2516-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR