Methyl-4-ethoxybenzoate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4-Ethoxybenzoic acid methyl ester, Methyl 4-ethoxybenzoate

M2738-25G M2738-5G
TCM2738-25GEA 149.35 USD
TCM2738-25G TCM2738-5G
Methyl-4-ethoxybenzoate ≥98.0% (by GC)
Methyl-4-ethoxybenzoate
Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Boiling Pt: 260°C
Storage Temperature: Refrigerator
MDL Number: MFCD00017264
CAS Number: 23676-08-6
EINECS: 245-817-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR