Methylprednisolone acetate

Supplier: TCI America

M2635-1G M2635-250MG
TCM2635-1GEA 222.48 USD
TCM2635-1G TCM2635-250MG
Methylprednisolone acetate
Methylprednisolone acetate
Formula: C₂₄H₃₂O₆
MDL Number: MFCD00200356
CAS Number: 53-36-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR