7-Iodopyrrolo[2,1-f][1,2,4]triazin-4-amine

Supplier: TCI America

I1139-1G I1139-5G
TCI1139-1GEA 99.91 USD
TCI1139-1G TCI1139-5G
7-Iodopyrrolo[2,1-f][1,2,4]triazin-4-amine
7-Iodopyrrolo[2,1-f][1,2,4]triazin-4-amine
Formula: C₆H₅IN₄
MDL Number: MFCD21607515
CAS Number: 1770840-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR