2-Iodo-4-methylphenol

Supplier: TCI America

I1132-1G I1132-5G
TCI1132-1GEA 61.8 USD
TCI1132-1G TCI1132-5G
2-Iodo-4-methylphenol
2-Iodo-4-methylphenol
Formula: C₇H₇IO
MDL Number: MFCD00189405
CAS Number: 16188-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR