4,5,6,7-Tetrachloro-2-hydroxyisoindoline-1,3-dione

Supplier: TCI America

Synonyms: N-Hydroxytetrachlorophthalimide

H1765-1G H1765-5G
TCH1765-1GEA 29.87 USD
TCH1765-1G TCH1765-5G
4,5,6,7-Tetrachloro-2-hydroxyisoindoline-1,3-dione
4,5,6,7-Tetrachloro-2-hydroxyisoindoline-1,3-dione
Formula: C₈HCl₄NO₃
MW: 300.91 g/mol
MDL Number: MFCD29088000
CAS Number: 85342-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR