2,3'-Bipyridine

Supplier: TCI America

D1772-1G D1772-5G
TCD1772-1GEA 24.72 USD
TCD1772-1G TCD1772-5G
2,3'-Bipyridine
2,3'-Bipyridine
Formula: C₁₀H₈N₂
MW: 156.184 g/mol
MDL Number: MFCD00006215
CAS Number: 581-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR