4-Amino-7-bromo-pyrrolo[2,1-f][1,2,4]triazine

Supplier: TCI America

Synonyms: 7-Bromopyrrolo[2,1-f][1,2,4]triazin-4-amine

B6237-1G B6237-5G
TCB6237-1GEA 57.68 USD
TCB6237-1G TCB6237-5G
4-Amino-7-bromo-pyrrolo[2,1-f][1,2,4]triazine
4-Amino-7-bromo-pyrrolo[2,1-f][1,2,4]triazine
Formula: C₆H₅BrN₄
MW: 213.04 g/mol
MDL Number: MFCD09033848
CAS Number: 937046-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR