Fmoc-Dap(Boc)-OH ≥97.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
Fmoc-(N-beta-boc)-L-alpha,beta-diaminopropionic acid, 3-(tert-Butoxycarbonyl)amino-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-alanine, (S)-3-(tert-Butoxycarbonylamino)-2-[(9H-fluoren-9-ylmethoxy)carbonylamino]propionic acid, Nα-Fmoc-Nβ-Boc-L-2,3-diaminopropionic acid, (S)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid, 3-(Boc-amino)-N-Fmoc-L-alanine
B6107-1G
B6107-5G
TCB6107-1GEA
105.06
USD
TCB6107-1G
TCB6107-5G
Fmoc-Dap(Boc)-OH ≥97.0% (by HPLC, titration analysis)
Fmoc-Dap(Boc)-OH
Formula:
C₂₃H₂₆N₂O₆ MW: 426.47 g/mol Melting Pt: 63°C Storage Temperature: Refrigerator |
MDL Number:
MFCD00235896 CAS Number: 162558-25-0 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Neutralization titration) | min. 97.0 % (calcd.on anh.substance) |
Specific rotation [a]20/D | -7.0 to -9.0 deg (C=, MeOH) (calcd.on anh.substance) |
Water | max. 3.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...