Benzyl (6-hydroxyhexyl)carbamate

Supplier: TCI America

Synonyms: 6-(N-Benzyloxycarbonylamino)-1-hexanol

B6075-1G B6075-5G
TCB6075-1GEA 77.25 USD
TCB6075-1G TCB6075-5G
Benzyl (6-hydroxyhexyl)carbamate
Benzyl (6-hydroxyhexyl)carbamate
Formula: C₁₄H₂₁NO₃
MW: 251.32 g/mol
MDL Number: MFCD00859036
CAS Number: 17996-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR