1H-Indazole-5-boronic acid, pinacol ester, N1-BOC protected

Supplier: TCI America

B5992-1G B5992-200MG
TCB5992-1GEA 163.77 USD
TCB5992-1G TCB5992-200MG
1H-Indazole-5-boronic acid, pinacol ester, N1-BOC protected
1H-Indazole-5-boronic acid, pinacol ester, N1-BOC protected
Formula: C₁₈H₂₅BN₂O₄
MDL Number: MFCD08437629
CAS Number: 864771-44-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR