DiBAC₄(3)

Supplier: TCI America

B5593-25MG B5593-5MG
TCB5593-25MGEA 235.87 USD
TCB5593-25MG TCB5593-5MG
DiBAC₄(3)
DiBAC₄(3)
Formula: C₂₇H₄₀N₄O₆
MDL Number: MFCD07437857
CAS Number: 70363-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR