1-n-Butyl-3-methylimidazolium nitrate

Supplier: TCI America

B5103-25G B5103-5G
TCB5103-25GEA 216.3 USD
TCB5103-25G TCB5103-5G
1-n-Butyl-3-methylimidazolium nitrate
1-n-Butyl-3-methylimidazolium nitrate
Formula: C₈H₁₅N₃O₃
CAS Number: 179075-88-8
UN: 1479

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR