7-Bromo-1-heptene

Supplier: TCI America

B3255-1G B3255-5G
TCB3255-1GEA 47.38 USD
TCB3255-1G TCB3255-5G
7-Bromo-1-heptene
7-Bromo-1-heptene
Formula: C₇H₁₃Br
MW: 177.085 g/mol
MDL Number: MFCD01631137
CAS Number: 4117-09-3
UN: 1993

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR