Sarcosine sodium salt ∼35% in water

Supplier: TCI America

Synonyms: Sodium Sarcosinate, N-Methylglycine Sodium Salt, Me-Gly-ONa, (Methylamino)acetic Acid Sodium Salt, Sodium (Methylamino)acetate

A3280-25ML A3280-100ML
TCA3280-25MLEA 59.74 USD
TCA3280-25ML TCA3280-100ML
Sarcosine sodium salt ∼35% in water
Sarcosine sodium salt
Formula: C₃H₇NO₂
MW: 111.08 g/mol
MDL Number: MFCD00068282
CAS Number: 4316-73-8
UN: 3267

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR