4-Amino-α,α,α-trifluoro-o-toluic acid

Supplier: TCI America

Synonyms: 4-Amino-2-(trifluoromethyl)benzoic acid

A2889-25G A2889-5G
TCA2889-25GEA 303.85 USD
TCA2889-25G TCA2889-5G
4-Amino-α,α,α-trifluoro-o-toluic acid
4-Amino-α,α,α-trifluoro-o-toluic acid
Formula: C₈H₆F₃NO₂
MDL Number: MFCD03407959
CAS Number: 393-06-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR