Neratinib

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: HKI-272, (E)-N-(4-((3-Chloro-4-(pyridin-2-ylmethoxy)phenyl)amino)-3-cyano-7-ethoxyquinolin-6-yl)-4-(dimethylamino)but-2-enamide

464700010 464702500
TS464700010EA 316 USD
TS464700010 TS464702500
Neratinib
Neratinib

  • Bioactive

Formula: C₃₀H₂₉ClN₆O₃
MW: 557 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09752958
CAS Number: 698387-09-6

Order Now

Specification Test Results

Appearance White to light yellow to beige Powder
Infrared spectrum Conforms
Solubility (1% in DMSO) Clear colorless to light yellow
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR