N6-Benzoyladenosine

Supplier: Thermo Scientific Chemicals
Warning

464690010 464690050
TS464690010EA 89 USD
TS464690010 TS464690050
N6-Benzoyladenosine
N6-Benzoyladenosine

  • Bioactive

Formula: C₁₇H₁₇N₅O₅
Melting Pt: 141…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00056988
CAS Number: 4546-55-8

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder or crystals
Infrared spectrum Conforms
Water ≤5 %
HPLC ≥95.0 %
Specific optical rotation -46° to -52° (20°C, 589 nm) (1% in DMSO)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR