β-L-Thymidine

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Telbivudine, 1-((2S,4R,5S)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione

464550010 464550050
TS464550010EA 120 USD
TS464550010 TS464550050
β-L-Thymidine
β-L-Thymidine

  • Bioactive

Formula: C₁₀H₁₄N₂O₅
MW: 242.2286 g/mol
Melting Pt: 185…190 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02683612
CAS Number: 3424-98-4

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Melting point 185 to 190 °C
Loss on drying ≤0.5 %
HPLC ≥98.5 %
Residue after ignition ≤0.2 %
Specific optical rotation -15° to -22° (20 °C, 589 nm) (1% in water)
Clarity of solution (1% in water) Clear colorless solution

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR