Carteolol hydrochloride

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-[3-[(1,1-Dimethylethyl)amino]-2-hydroxypropoxy]-3,4-dihydro-2(1H)-quinolinone hydrochloride

464320010 464320050
TS464320010EA 128 USD
TS464320010 TS464320050
Carteolol hydrochloride
Carteolol hydrochloride

  • Bioactive

Formula: C₁₆H₂₄N₂O₃·HCl
Melting Pt: 278 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00941499
CAS Number: 51781-21-6

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (1 g, 105°C)
Solubility (0.3 g in 10 ml water) Clear colorless solution
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR