Ibandronate sodium salt monohydrate

Supplier: Thermo Scientific Chemicals

463150010 463150050
TS463150010EA 129 USD
TS463150010 TS463150050
Ibandronate sodium salt monohydrate
Ibandronate sodium salt monohydrate

  • Bioactive

Formula: C₉H₂₂NNaO₇P₂·H₂O
Melting Pt: 175…177 °C
Storage Temperature: Freezer
MDL Number: MFCD07197214
CAS Number: 138926-19-9

Order Now

Specification Test Results

Appearance White to almost white Powder
Infrared spectrum Conforms
Loss on drying 4.5 % to 8.5 %
Titration with NaOH 96 to 104 % (on dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR