Cefditoren pivoxil

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (6R,7R)-(pivaloyloxy)methyl 7-((Z)-2-(2-aminothiazol-4-yl)-2-(methoxyimino)acetamido)-3-((Z)-2-(4-methylthiazol-5-yl)vinyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate

463110010 463110050
TS463110010EA 125 USD
TS463110010 TS463110050
Cefditoren pivoxil
Cefditoren pivoxil

  • Bioactive

Formula: C₂₅H₂₈N₆O₇S₃
MW: 620.71 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00933166
CAS Number: 117467-28-4

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Water ≤1.5 %
Specific optical rotation -45° to -52° (20°C, 589 nm) (0.5% in MeOH)
Titration with HClO4 94.0 to 106.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR