Risedronic acid sodium salt

Supplier: Thermo Scientific Chemicals
Warning

462930010 462932500
TS462930010EA 321 USD
TS462930010 TS462932500
Risedronic acid sodium salt
Risedronic acid sodium salt

  • Bioactive

Formula: C₇H₁₀NNaO₇P₂
Storage Temperature: Refrigerator
MDL Number: MFCD01706268
CAS Number: 115436-72-1

Order Now

Specification Test Results

Appearance White to beige Powder
Infrared spectrum Conforms
Water 11.9 to 13.9 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR