5-Fluoro-5'-deoxyuridine (Doxifluridine)

Supplier: Thermo Scientific Chemicals

Synonyms: (+)-5-Fluoro-5'-deoxyuridine, 1-(5-Deoxy-beta-D-ribofuranosyl)-5-fluorouracil, Doxifluridine, 5'-Deoxy-5-fluorouridine, 5'-dFUrd, 1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-methyltetrahydrofuran-2-yl)-5-fluoropyrimidine-2,4(1H,3H)-dione

462790010 462790050
TS462790010EA 77 USD
TS462790010 TS462790050
5-Fluoro-5'-deoxyuridine (Doxifluridine)
5-Fluoro-5'-deoxyuridine (Doxifluridine)

  • Bioactive

Formula: C₉H₁₁FN₂O₅
MW: 246.19 g/mol
Melting Pt: 188…192 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00866530
CAS Number: 3094-09-5

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Melting point 187 to 193 °C
Loss on drying ≤0.5 %
HPLC ≥97.5 %
Specific optical rotation +18° to +22° (20 °C, 589 nm) (1 % in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR