1,2-Bis(trichlorosilyl)ethane

Supplier: Thermo Scientific Chemicals
Danger

462030050 462030250
TS462030050EA 81 USD
TS462030050 TS462030250
1,2-Bis(trichlorosilyl)ethane
1,2-Bis(trichlorosilyl)ethane

  • Bioactive

Formula: C₂H₄Cl₆Si₂
MW: 296.92 g/mol
Melting Pt: 24…25 °C
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00013601
CAS Number: 2504-64-5
UN: 2987
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to light yellow Crystalline mass or liquid
Infrared spectrum Conforms
Refractive index 1.4735 to 1.4760 (20°C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR