Khellin ≥95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Visammin, 4,9-Dimethoxy-7-methyl-5H-furo[3,2-g]chromen-5-one, 4,9-Dimethoxy-7-methyl-5H-furo[3,2-g][1]benzopyran-5-one

461800050 461800250
TS461800050EA 95 USD
TS461800050 TS461800250
Khellin ≥95%
Khellin

  • Bioactive

Formula: C₁₄H₁₂O₅
MW: 260.25 g/mol
Melting Pt: 150…154 °C
Storage Temperature: Ambient
MDL Number: MFCD00005007
CAS Number: 82-02-0
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Light yellow to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 150 to 154 °C
Water ≤1 %
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR