Caffeine citrate

Supplier: Thermo Scientific Chemicals
Warning

461720250 461721000
TS461720250EA 121 USD
TS461720250 TS461721000
Caffeine citrate
Caffeine citrate
Formula: C₁₄H₁₈N₄O₉
Melting Pt: 159…161 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00044986
CAS Number: 69-22-7

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤5 %
Titration with HClO4 48 to 52 % (Caffeine)
Titration with TBAH 48 to 52 % (Citric acid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR