Isoconazole nitrate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-[2-(2,6-Dichlorobenzyloxy)-2-(2,4-dichlorophenyl)ethyl]imidazole Nitrate, 1-(2-((2,6-Dichlorobenzyl)oxy)-2-(2,4-dichlorophenyl)ethyl)-1H-imidazole nitrate

458640050 458640250 458640010
TS458640050EA 71 USD
TS458640050 TS458640250 TS458640010
Isoconazole nitrate 99%
Isoconazole nitrate
Formula: C₁₈H₁₄Cl₄N₂O
MW: 479.14 g/mol
Melting Pt: 181…183 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01721647
CAS Number: 24168-96-5

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 178 to 182 °C
Titration with HClO4 ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR