Bifonazole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(alpha,4-Diphenylbenzyl)imidazole, 1-([1,1'-Biphenyl]-4-yl(phenyl)methyl)-1H-imidazole

458620010 458620050 458620250
TS458620010EA 28 USD
TS458620010 TS458620050 TS458620250
Bifonazole 98%
Bifonazole
Formula: C₂₂H₁₈N₂
Melting Pt: 149 °C
Storage Temperature: Ambient
MDL Number: MFCD00865567
CAS Number: 60628-96-8

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 148 to 153 °C
Loss on drying ≤0.5 % (1 g, 105 °C, 1 h)
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR