Polyvinyl alcohol, M.W. ∼ 13,000 98%, hydrolysed

Supplier: Thermo Scientific Chemicals

Synonyms: Poly(vinyl alcohol), PVOH, PVA, Mowiol®

458240250 458241000 458245000
TS458240250EA 42 USD
TS458240250 TS458241000 TS458245000
Polyvinyl alcohol, M.W. ∼ 13,000 98%, hydrolysed
Polyvinyl alcohol
Formula: [-CH₂CH(OH)-]n
Melting Pt: 230…240 °C
Storage Temperature: Ambient
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Order Now

Specification Test Results

Appearance Off-white to light yellow Powder or crystals
Infrared spectrum Conforms
Assay 98 to 98.8 mol % (degree of hydrolysis)
Viscosity 3.5 to 4.5 cP (4 % in water)
Ash ≤1.2 % (as Na2O)
pH 5.0 to 7.0 (4 % in water)
Volatile matter ≤5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR