3,4-Dihydroisoquinolin-1(2H)-one 98%
Supplier: Thermo Scientific Chemicals
458220010
458220050
TS458220010EA
124
USD
TS458220010
TS458220050
3,4-Dihydroisoquinolin-1(2H)-one 98%
3,4-Dihydroisoquinolin-1(2H)-one
Formula:
C9H9NO Melting Pt: 67…70 °C Storage Temperature: Ambient |
MDL Number:
MFCD00853963 CAS Number: 1196-38-9 |
Specification Test Results
Appearance | White to yellow Powder or crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...