Sulfachloropyridazine sodium 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Sodium sulphachloropyridazine, Prinzone

458060010 458060050
TS458060010EA 94 USD
TS458060010 TS458060050
Sulfachloropyridazine sodium 97%
Sulfachloropyridazine sodium
Formula: C₁₀H₈ClN₄NaO₂S
MW: 306.70 g/mol
Melting Pt: 262…270 °C
Storage Temperature: Ambient
MDL Number: MFCD04117866
CAS Number: 23282-55-5

Order Now

Specification Test Results

Appearance Off-white to beige Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤2 %
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR