1-Chloromethyl-1H-1,2,4-triazole hydrochloride 98%

Supplier: Thermo Scientific Chemicals

456090010 456090050
TS456090010EA 63 USD
TS456090010 TS456090050
1-Chloromethyl-1H-1,2,4-triazole hydrochloride 98%
1-Chloromethyl-1H-1,2,4-triazole hydrochloride
Formula: C₃H₄ClN₃·HCl
MW: 154.00 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00964947
CAS Number: 84387-61-1

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR