2-Chloro-4-formylpyridine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Chloropyridine-4-carboxaldehyde, 2-Chloroisonicotinaldehyde
456040050
456040010
TS456040050EA
162
USD
TS456040050
TS456040010
2-Chloro-4-formylpyridine 97%
2-Chloro-4-formylpyridine
Formula:
C₆H₄ClNO MW: 141.56 g/mol Melting Pt: 46…47 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD06651557 CAS Number: 101066-61-9 |
Specification Test Results
Appearance | White to yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 44 to 50 °C |
Water | ≤1.5 % |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...