6-(Hydroxymethyl)indole 97%

Supplier: Thermo Scientific Chemicals
Warning

455540010 455542500
TS455540010EA 277 USD
TS455540010 TS455542500
6-(Hydroxymethyl)indole 97%
6-(Hydroxymethyl)indole
Formula: C₉H₉NO
Melting Pt: 45…50 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02179595
CAS Number: 1075-26-9

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 60 to 67 °C
Water ≤1 %
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR