Methyl-3-cyclohexene-1-carboxylate 97%

Supplier: Thermo Scientific Chemicals
Warning

452860250 452861000
TS452860250EA 29 USD
TS452860250 TS452861000
Methyl-3-cyclohexene-1-carboxylate 97%
Methyl-3-cyclohexene-1-carboxylate
Formula: C₈H₁₂O₂
Boiling Pt: 185 °C
Density: 1.02 g/cm³
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00053266
CAS Number: 6493-77-2
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4600 to 1.4620 (20 °C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR