DL-Valine 98%, Extra Pure

Supplier: Thermo Scientific Chemicals

452830010 452832500
TS452830010EA 443 USD
TS452830010 TS452832500
DL-Valine 98%, Extra Pure
DL-Valine
Formula: C₅H₁₁NO₂
Melting Pt: 283.5…285 °C (dec.)
Density: 1.31 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00004267
CAS Number: 516-06-3

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Water ≤2 %
Titration with HClO4 ≥97.5 %
Heavy metals (as Pb) ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR