3,4-Dimethylthiophene 97%

Supplier: Thermo Scientific Chemicals
Warning

450950010 450950050
TS450950010EA 141 USD
TS450950010 TS450950050
3,4-Dimethylthiophene 97%
3,4-Dimethylthiophene
Formula: C₆H₈S
Boiling Pt: 144 °C (760 mmHg)
Flash Pt: 26 °C
Storage Temperature: Ambient
MDL Number: MFCD01764731
CAS Number: 632-15-5
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow (may darken on storage) Liquid
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR