7-Chloro-6-nitro-4-hydroxyquinazoline 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 7-Chloro-6-nitroquinazolin-4(3H)-one, 7-Chloro-6-nitro-4(3H)-quinazolinone, 7-Chloro-6-nitro-4-quinazolinol, 7-Chloro-6-nitro-4-quinazolinone

450790050 450790250
TS450790050EA 31 USD
TS450790050 TS450790250
7-Chloro-6-nitro-4-hydroxyquinazoline 97%
7-Chloro-6-nitro-4-hydroxyquinazoline
Formula: C₈H₄ClN₃O₃
MW: 225.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09388771
CAS Number: 53449-14-2
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to yellow to brown Crystalline powder
Infrared spectrum Conforms
Water ≤0.5 %
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR