N-Ethyl-p-toluidine 97%

Supplier: Thermo Scientific Chemicals
Danger

450780250 450781000
TS450780250EA 66 USD
TS450780250 TS450781000
N-Ethyl-p-toluidine 97%
N-Ethyl-p-toluidine
Formula: C₉H₁₃N
Boiling Pt: 218 °C
Density: 0.938 g/cm³
Flash Pt: 89 °C
Storage Temperature: Ambient
MDL Number: MFCD00035793
CAS Number: 622-57-1
UN: 2754
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5435 to 1.5455 (20 °C, 589 nm)
Water ≤0.5 %
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR