Methyl-3-amino-4-methylthiophene-2-carboxylate 98%

Supplier: Thermo Scientific Chemicals
Warning

450770050 450770250
TS450770050EA 43 USD
TS450770050 TS450770250
Methyl-3-amino-4-methylthiophene-2-carboxylate 98%
Formula: C₇H₉NO₂S
Melting Pt: 85…88 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00051822
CAS Number: 85006-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR