5-Methylpyrimidine 98%

Supplier: Thermo Scientific Chemicals
Warning

450730010 450732500
TS450730010EA 126 USD
TS450730010 TS450732500
5-Methylpyrimidine 98%
5-Methylpyrimidine
Formula: C₅H₆N₂
Boiling Pt: 152…154 °C
Melting Pt: 33…37 °C
Flash Pt: 52 °C
Storage Temperature: Ambient
MDL Number: MFCD00047386
CAS Number: 2036-41-1
UN: 1325
ADR: 4.1,III

Order Now

Specification Test Results

Appearance White to tan Crystalline powder or crystals or solid
Infrared spectrum Conforms
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR