1-Phenacylpyridinium bromide 97%

Supplier: Thermo Scientific Chemicals
Warning

450120100 450120500
TS450120100EA 20 USD
TS450120100 TS450120500
1-Phenacylpyridinium bromide 97%
1-Phenacylpyridinium bromide
Formula: C₁₃H₁₂BrNO
Melting Pt: 193…195 °C
Storage Temperature: Ambient
MDL Number: MFCD00031995
CAS Number: 16883-69-5

Order Now

Specification Test Results

Appearance Off-white to light beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 196 to 200 °C
Titration Argentometric ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR