Ethyl-4-chloro-8-cyanoquinoline-3-carboxylate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 4-chloro-8-cyanoquinoline-3-carboxylate

449680010 449680050
TS449680010EA 142 USD
TS449680010 TS449680050
Ethyl-4-chloro-8-cyanoquinoline-3-carboxylate 97%
Ethyl 4-chloro-8-cyanoquinoline-3-carboxylate
Formula: C₁₃H₉ClN₂O₂
MW: 260.68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00173414
CAS Number: 77173-67-2

Order Now

Specification Test Results

Appearance Light yellow to light brown Powder
Infrared spectrum Conforms
Water ≤1 %
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR