5-Nitro-2-thiophenecarbonitrile 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 5-Nitrothiophene-2-carbonitrile

449370010 449370050
TS449370010EA 36 USD
TS449370010 TS449370050
5-Nitro-2-thiophenecarbonitrile 98%
5-Nitro-2-thiophenecarbonitrile
Formula: C₅H₂N₂O₂S
MW: 154.15 g/mol
Melting Pt: 42…46 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00052401
CAS Number: 16689-02-4
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Yellow to beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 42 to 46 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR