4'-Isobutylacetophenone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: p-Isobutylacetophenone

448820500 448822500
TS448820500EA 81 USD
TS448820500 TS448822500
4'-Isobutylacetophenone 97%
4'-Isobutylacetophenone
Formula: C₁₂H₁₆O
MW: 176.26 g/mol
Boiling Pt: 107 °C (2 mmHg)
Density: 0.96 g/cm³
Flash Pt: 54 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00027393
CAS Number: 38861-78-8
UN: 1224
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5170 to 1.5190 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR