4-Chloro-m-anisidine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 5-Amino-2-chloroanisole, 4-Chloro-3-methoxyaniline, 4-Chloro-3-anisidine

448800010 448800050
TS448800010EA 17 USD
TS448800010 TS448800050
4-Chloro-m-anisidine 98%
4-Chloro-m-anisidine
Formula: C₇H₈ClNO
MW: 157.6 g/mol
Melting Pt: 78…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00672967
CAS Number: 13726-14-2
UN: 2233
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 78 to 81 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR