2,6-Dimethoxyaniline 96%

Supplier: Thermo Scientific Chemicals
Danger

448790010 448790050
TS448790010EA 71 USD
TS448790010 TS448790050
2,6-Dimethoxyaniline 96%
2,6-Dimethoxyaniline
Formula: C₈H₁₁NO₂
MW: 153.18 g/mol
Melting Pt: 76…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00053934
CAS Number: 2734-70-5
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Grey to brown Crystalline powder
Infrared spectrum Conforms
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR